Learn More
Research Products International Corp Shorty 25 ml Serological Pipets, Bulk Packed, Sterile, 10 per Bag, 100 per Package
SDP
Catalog No. 50998782
Encompass
The Shorty Serological Pipet features an exclusive one piece molded design that offers unique advantages compared to standard welded tip pipets. The pipet barrel is almost two inches shorter to allow for easier and convenient handling under tissue culture hoods and benchtop enclosures. One piece style features printed reference line marks. Manufactured from high grade polystyrene with standard tip opening for cell culture applications.
Catalog No. 50-998-782 Supplier Research Products International Corp Supplier No. 248606
The Shorty Serological Pipet features an exclusive one piece molded design that offers unique advantages compared to standard welded tip pipets. The pipet barrel is almost two inches shorter to allow for easier and convenient handling under tissue culture hoods and benchtop enclosures. One piece style features printed reference line marks. Manufactured from high grade polystyrene with standard tip opening for cell culture applications.