Learn More
CELLTREAT Scientific Products 25mL Serological Serological Pipet , Individual Paper/Plastic Wrapper Packed in Cartons, Sterile pipette
SDP
Catalog No. 50202118
Encompass_Preferred
CELLTREAT Scientific Products 25 Serological Pipet. Convenient shelf carton allows easy access to pipets and neat storage. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. RNase/DNase Free, Non-pyrogenic. Every case is is printed with the lot number for quality traceability. Packaged sterile, 50 individually wrapped in paper/plastic wrapper per dispensor carton, 200/case.
Catalog No. 50-202-118 Supplier CELLTREAT Scientific Products Supplier No. 229225
CELLTREAT Scientific Products 25 Serological Pipet. Convenient shelf carton allows easy access to pipets and neat storage. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. RNase/DNase Free, Non-pyrogenic. Every case is is printed with the lot number for quality traceability. Packaged sterile, 50 individually wrapped in paper/plastic wrapper per dispensor carton, 200/case.