Learn More
CELLTREAT Scientific Products 25mL Serological Pipet, Bulk Packed in Bags, Sterile pipette
SDP
Supplier:  CELLTREAT Scientific Products 229245
Encompass_Preferred
CELLTREAT Scientific Products 25mL Serological Pipet. Convenient bulk packaging option. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. -10mL graduations. Every case is printed with the lot number for quality traceability. Packaged sterile, 25 bulk packed/re-sealable bag, 200/case.
Catalog No. 50-202-124