Learn More
Cole-Parmer Cole-Parmer Essentials Pipette Basins, 25 mL, Sterile, Individually Wrapped; 50/cs
Catalog No. NC1986435 Shop All Cole-Parmer Products
Encompass_Preferred
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.
This item is not returnable. View return policy
Catalog No. NC1986435 Supplier Cole-Parmer Supplier No. 0439523
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.