Learn More
CELLTREAT Scientific Products 25mL Suspension Culture Flask - Vent Cap, Sterile
SDP
Catalog No. 50202083
Encompass_Preferred
CELLTREAT Scientific Products 25mL Suspension Culture Flask - Non-treated. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Vent caps feature 0.22¿m hydrophobic membrane. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 200/case.
This item is not returnable. View return policy
Catalog No. 50-202-083 Supplier CELLTREAT Scientific Products Supplier No. 229500
CELLTREAT Scientific Products 25mL Suspension Culture Flask - Non-treated. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Vent caps feature 0.22¿m hydrophobic membrane. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 200/case.