Learn More
Hach Company 25 mL Glass Syringe for AT/KF Titrator
Catalog No. NC3368694 Shop All Hach Company Products
Encompass
Glass 25 mL syringe designed for use with AT/KF titrators to accurately deliver and withdraw titrant during Karl Fischer and automatic titration procedures.
  • Compatible with AT/KF titrators.
  • 25 mL capacity for titration operations.
  • Glass construction suitable for titration applications.
Catalog No. NC3368694 Supplier Hach Company Supplier No. LZE103
Glass 25 mL syringe designed for use with AT/KF titrators to accurately deliver and withdraw titrant during Karl Fischer and automatic titration procedures.
  • Compatible with AT/KF titrators.
  • 25 mL capacity for titration operations.
  • Glass construction suitable for titration applications.