Learn More
Lab Products Inc 25ML VIAL CLR 28X70MM/149CS
Catalog No. NC9785493
Encompass_Preferred
Vial 25 Ml Clear Glass Screw Thread 28X70Mm 149/Case
Catalog No. NC9785493 Supplier Lab Products Inc Supplier No. 325C1441
Vial 25 Ml Clear Glass Screw Thread 28X70Mm 149/Case