Learn More
Beckman Coulter Syringe, 25 mL - Replacement for Beckman Coulter HIAC 9703+ (690-300-4809)
Catalog No. NC2621509
Encompass
Replacement 25 mL syringe for use with Beckman Coulter HIAC liquid particle counters. Listed as an accessory in the HIAC 9703+ user manual, it serves as a consumable for sample aspiration and controlled fluid delivery in particle counting workflows.
  • 25 mL capacity.
  • Replacement consumable for HIAC liquid particle counters.
  • Used for precise sample aspiration and controlled fluid delivery.
  • Helps maintain instrument performance and reduce downtime.
Catalog No. NC2621509 Supplier Beckman Coulter Supplier No. 6903004809
Replacement 25 mL syringe for use with Beckman Coulter HIAC liquid particle counters. Listed as an accessory in the HIAC 9703+ user manual, it serves as a consumable for sample aspiration and controlled fluid delivery in particle counting workflows.
  • 25 mL capacity.
  • Replacement consumable for HIAC liquid particle counters.
  • Used for precise sample aspiration and controlled fluid delivery.
  • Helps maintain instrument performance and reduce downtime.