Learn More
Ace Glass, Inc. 25mm Ace-Thred nylon bushing, -214 CAPFE O-ring, accepts24-25mm OD tubing
SDP
Catalog No. 50974963
Encompass_Preferred
Catalog No. 50-974-963 Supplier Ace Glass, Inc. Supplier No. 750611