Learn More
Ace Glass, Inc. 25MM 4MM BORE ADAPTER 25MM 4MM BORE ADAPTER
SDP
Supplier:  Ace Glass, Inc. 583517
Encompass_Preferred
25mm adapter with drip tip for chemical type chromatography. Adapter has 4mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.
Catalog No. 50-972-233