Learn More
Cytiva Acrodisc Syringe Filter with wwPTFE Membrane - 0.2 µm, 25 mm (1000/pkg)
Supplier:  Cytiva 4929
Encompass
Acrodisc Syringe Filter with wwPTFE Membrane - 0.2 µm, Standard 25 mm.Syringe filters with wwPTFE membrane are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 μm) and UHPLC (0.2 μm) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime. The syringe filters are low in protein and API binding which make them ideal for drug studies, such as dissolution testing. Both the membrane and housing are extremely low in levels of potential extractables.25 mm Standard Acrodisc Syringe Filters are ideal for sample volumes of ≤ 100 mL and feature an effective filtration area of 2.8 cm2. They feature a polypropylene housing that is chemically resistant to harsh solvents and low in extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3124