Learn More
Ace Glass, Inc. 25MM ADAPTER 25MM ADAPTER
SDP
Catalog No. 50973331
Encompass_Preferred
25mm O.D. screen adapter used with ACE freeze drying vessels for assuring that the contents of the vessel do not accidentally get sucked out during the freeze drying process. Adapter is PTFE with a stainless steel screen, and press fits into the end of the vessel before attaching it to the Ace-Thred.
Catalog No. 50-973-331 Supplier Ace Glass, Inc. Supplier No. 669684
25mm O.D. screen adapter used with ACE freeze drying vessels for assuring that the contents of the vessel do not accidentally get sucked out during the freeze drying process. Adapter is PTFE with a stainless steel screen, and press fits into the end of the vessel before attaching it to the Ace-Thred.