Learn More
Electron Microscopy Sciences Air Sampling 25 mm Asbestos PCM Cassette with Gridded Filter
SDP
Supplier:  Electron Microscopy Sciences 6521450
Encompass
PCM 25 mm air sampling cassettes conform to all NIOSH, OSHA, and EPA standards. The cassette housings are injection molded in-house and are individually assembled and preloaded under stringent quality control standards. Once assembled, each cassette is individually tested prior to packaging for shipment. Random lot sample testing is performed daily by an independent laboratory to ensure low background fiber counts. Laboratory results are available upon request. Quality 0.8 um MCE filter material; 2" Static conductive extension cowl; Meets NIOSH 7400 specifications; Preloaded - ready-to-use. Grid lines printed on filter.
Catalog No. 50-338-51