Learn More
Electron Microscopy Sciences Air Sampling 25 mm Asbestos PCM Cassette with Green Gridded Filter
SDP
Supplier:  Electron Microscopy Sciences 6521550
Encompass
This PCM 25 mm air sampling cassettes are designed and manufactured to conform to all NIOSH, OSHA, and EPA standards. The cassette housings are injection molded in-house and are individually assembled and preloaded under stringent quality control standards. Once assembled, each cassette is individually tested prior to packaging for shipment. Random lot sample testing is performed daily by an independent laboratory to ensure low background fiber counts. Laboratory results are available upon request.
Features
Quality 0.8µm MCE filter material
2" Static conductive extension cowl
Meets NIOSH 7400 specifications
Preloaded – ready-to-use
Green Grid lines printed on filter
Catalog No. 50-338-52