Learn More
Azzota Corp Disposable Microporous Filter, Nylon Membrane, 25mm Diameter, Plain Surface, White, 0.45M Pore Size. 200 Pack
Catalog No. 507532922
Encompass_Preferred
Disposable Microporous Filter, NYLON Membrane, 25mm Diameter, plain Surface, White, 0.45m Pore Size. 200 pack
Catalog No. 50-753-2922 Supplier Azzota Corp Supplier No. BF250145
Disposable Microporous Filter, NYLON Membrane, 25mm Diameter, plain Surface, White, 0.45m Pore Size. 200 pack