Learn More
Azzota Corp Disposable Syringe Filter, Ptfe Membrane, High Density Polypropylene (Pp) Housing, 0.45M Pore Size, Filter Diameter 25mm, Pack Of 100
Supplier:  Azzota Corp AT250145
Encompass_Preferred
Disposable Syringe Filter, PTFE Membrane, High density Polypropylene (PP) Housing, 0.45m Pore Size, Filter diameter 25mm, Pack of 100
Catalog No. 50-753-2937