Learn More
Chemglass Life Sciences #25 Quartz O-Ring Joint, 28.8mm Tube O.D., Clamp Size/35, 217 O-Ring Size
SDP
Catalog No. CGQ013507
Encompass_Preferred
#25 Quartz O-Ring Joint, 28.8mm Tube O.D., Clamp Size/35, 217 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.
Catalog No. CGQ013507 Supplier Chemglass Life Sciences Supplier No. CGQ013507
#25 Quartz O-Ring Joint, 28.8mm Tube O.D., Clamp Size/35, 217 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.