Learn More
EZKEM Polypropylene Membrane for Gas Diffusion, 25 mm, Used for Cyanide or Ammonia/ TKN analysis.(10 Pack)
SDP
Catalog No. NC2980254
Encompass_Preferred
EZkem’s 25 mm Polypropylene Gas Diffusion Membrane is used for Cyanide or Ammonia/ TKN analysis in the gas diffusion module. Pack of 10.
Catalog No. NC2980254 Supplier EZKEM Supplier No. A001520
EZkem’s 25 mm Polypropylene Gas Diffusion Membrane is used for Cyanide or Ammonia/ TKN analysis in the gas diffusion module. Pack of 10.