Learn More
Ace Glass, Inc. 25MM GRADED SEALS 25MM GRADED SEALS
SDP
Catalog No. 50974602
Encompass
25mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-602 Supplier Ace Glass, Inc. Supplier No. 845536
25mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.