Learn More
Teknova 25mM Histidine, pH 6.0, 1000mL, Endotoxin Tested, Sterile. 8 pack.
SDP
Catalog No. NC2741155
Encompass
Amino Acids are essential nutrients for cell growth, and thus they are often used in the preparation of growth media. Amino acid solution can help to increase cell viability and growth rate, making it an essential component for cell culture.
Catalog No. NC2741155 Supplier Teknova Supplier No. H25608PK
Amino Acids are essential nutrients for cell growth, and thus they are often used in the preparation of growth media. Amino acid solution can help to increase cell viability and growth rate, making it an essential component for cell culture.