Learn More
As One International Inc 25 mM MgCl2, 10 x 1.5 ml
Catalog No. 501655984
Encompass_Preferred
25mM magnesium is for optimization of magnesium concentrations in PCR reactions. Optimal magnesium concentration increases fidelity and specificity of the polymerase. Too low of a concentration inactivates the polymerase while too high of a concentration increase nonspecific bands. Several variables effect the optimal concentration of magnesium, DNA quality, presence of chelators, and dNTP concentration.
Catalog No. 50-165-5984 Supplier As One International Inc Supplier No. AO308110
25mM magnesium is for optimization of magnesium concentrations in PCR reactions. Optimal magnesium concentration increases fidelity and specificity of the polymerase. Too low of a concentration inactivates the polymerase while too high of a concentration increase nonspecific bands. Several variables effect the optimal concentration of magnesium, DNA quality, presence of chelators, and dNTP concentration.