Learn More
Cytiva Acrodisc Syringe Filters with Nylon Membrane - 0.2 µm, 25mm (50/pkg 200/cs)
Supplier:  Cytiva 4436
Encompass
Acrodisc Syringe Filters with Nylon Membrane - 0.2 µm, Standard 25mm.Syringe filters with nylon membrane are ideal for the filtration of aqueous and solvent-based sample prep for HPLC (0.45 μm) and UHPLC (0.2 μm) applications. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime.Nylon syringe filters show excellent chemical compatibility with esters, bases and alcohols. Nylon syringe filters are not recommended for use with strongly acidic fluids.25 mm Standard Acrodisc Syringe Filters are ideal for sample volumes of ≤ 100 mL and feature an effective filtration area of 2.8 cm2. They feature a polypropylene housing that is chemically resistant to harsh solvents and low in extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1476514