Learn More
Ace Glass, Inc. Coupling, #25 to #25 Ace-Thred™;, nylon, front seal FETFE; O-rings
SDP
Catalog No. 50978501
Encompass_Preferred
25mm diameter thread (#25 Ace-Thred) nylon coupling. For coupling threaded columns together, or coupling threaded columns to end fittings, with single o-ring seal for leak-tight engagement. Supplied with two FETFE o-rings.
Catalog No. 50-978-501 Supplier Ace Glass, Inc. Supplier No. 584116
25mm diameter thread (#25 Ace-Thred) nylon coupling. For coupling threaded columns together, or coupling threaded columns to end fittings, with single o-ring seal for leak-tight engagement. Supplied with two FETFE o-rings.