Learn More
Ace Glass, Inc. Fritted disc, 25mm OD, porosity A (145-174 micron), fits3700-10
SDP
Catalog No. 50978188
Encompass_Preferred
25mm O.D. Porosity A (145-174 micron) fritted disc.
Catalog No. 50-978-188 Supplier Ace Glass, Inc. Supplier No. 370323
25mm O.D. Porosity A (145-174 micron) fritted disc.