Learn More
Ace Glass, Inc. #25 Ace-Thred solid PTFE plug, 7855-734 FETFE O-ring front seal
SDP
Catalog No. 509557166
Encompass_Preferred
#25 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -212 FETFE O-ring.
Catalog No. 50-955-7166 Supplier Ace Glass, Inc. Supplier No. 584650
#25 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -212 FETFE O-ring.