Learn More
Chemglass Life Sciences 25mm Quartz Fritted Disc, Medium
SDP
Catalog No. CGQ020710
Encompass_Preferred
25mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020710 Supplier Chemglass Life Sciences Supplier No. CGQ020710
25mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.