Learn More
Ace Glass, Inc. 25mm x 51cm chromatography column
SDP
Catalog No. 50972562
Encompass_Preferred
25mm x 51cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring. Complete with ACE 5853-07 tubing connector for connecting 12.5 t0 14mm O.D. tubing to column.
Catalog No. 50-972-562 Supplier Ace Glass, Inc. Supplier No. 590245
25mm x 51cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring. Complete with ACE 5853-07 tubing connector for connecting 12.5 t0 14mm O.D. tubing to column.