Learn More
Ace Glass, Inc. 25mm x 51cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 590523
Encompass_Preferred
25mm X 51cm chromatography column with 2mm bore, 1:5 taper solid PTFE stopcock plug, porosity B fritted disc sealed in at bottom, and a #15 Ace-Thred at top of column.
Catalog No. 50-972-572