Learn More
Drg International Inc 25Oh Vitamin D Total Elisa Ruo
Supplier:  Drg International Inc EIA5396R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of total 25-OH Vitamin D (Vitamin D2/D3) in serum or plasma (EDTA, lithium heparin or citrate plasma). This product is for Research Use Only. The DRG 25-OH Vitamin D (total) ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the principle of competitive binding. The microtiter wells are coated with a monoclonal (mouse) antibody directed towards a unique antigenic site of 25-OH Vitamin D (25-OH D) molecule.
Catalog No. 50-194-7628