Learn More
Cayman Chemical 25P-NBOMehydrochlorIde 50mg
SDP
Catalog No. 502894671
Encompass_Preferred
A phenethylamine that binds 5-HT2A and 5-HT2C with similar affinities (pKis = ~9 for both)
Catalog No. 50-289-4671 Supplier Cayman Chemical Supplier No. 900199150 MG
A phenethylamine that binds 5-HT2A and 5-HT2C with similar affinities (pKis = ~9 for both)