Learn More
Medchemexpress LLC (25R)-Spirost-4-ene-3,12-dione | 6875-60-1 | 5 MG
SDP
Catalog No. 5000419081
Encompass_Preferred
(25R)-Spirost-4-ene-3,12-dione is a natural product that inhibits neutrophil superoxide anion production and histamine release from mast cells. It is for research use only and not sold to patients. This steroid has a molecular weight of 426.59, a chemical formula of C27H38O4, and appears as a white to off-white solid.
  • Inhibits neutrophil superoxide anion production
  • Inhibits histamine release from mast cells
  • Natural product
  • Classified as a steroid
Catalog No. 50-004-19081 Supplier Medchemexpress LLC Supplier No. HYN78855MG
(25R)-Spirost-4-ene-3,12-dione is a natural product that inhibits neutrophil superoxide anion production and histamine release from mast cells. It is for research use only and not sold to patients. This steroid has a molecular weight of 426.59, a chemical formula of C27H38O4, and appears as a white to off-white solid.
  • Inhibits neutrophil superoxide anion production
  • Inhibits histamine release from mast cells
  • Natural product
  • Classified as a steroid