Learn More
Cayman Chemical 25S-7-DafachronIc AcId 100ug
SDP
Catalog No. 502955489
Encompass_Preferred
A sterol-derived hormone that acts as a ligand of DAF-12 (EC50 = 23 nM; 50 nM, inhibits the dauer-promoting activity of DAF-12, blocking the formation of infective larvae in several parasitic nematodes in favor of reproductive maturation to adults
Catalog No. 50-295-5489 Supplier Cayman Chemical Supplier No. 14101100 UG
A sterol-derived hormone that acts as a ligand of DAF-12 (EC50 = 23 nM; 50 nM, inhibits the dauer-promoting activity of DAF-12, blocking the formation of infective larvae in several parasitic nematodes in favor of reproductive maturation to adults