Learn More
AdipoGen 25S-delta7-Dafachronic acid
SDP
Supplier:  AdipoGen AGCN20014C100
Encompass_Preferred
Chemical. CAS 949004-12-0. Formula C27H42O3. MW 414.6. Ligand of DAF-12, an orphan nuclear receptor, that regulates dauer diapause, reproductive development, fat metabolism, and life span life cycle in parasitic nematodes. Sterol-derived hormone. Blocks formation of infective larvae in several parasitic nematodes. Inhibits the dauer-promoting activity of DAF-12. Supports the reproductive development.
Catalog No. 50-256-2694