Learn More
Enzo Life Sciences Cathepsin B (human liver), (purified) (25 µg)
SDP
Catalog No. NC9990393
Encompass_Preferred
Highly pure lysosomal protease. MW: ~27.5 kDa. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM sodium acetate, pH 5.0, containing 1mM EDTA. Source: Purified from human liver. UniProt: P07858. Long Term Storage: -80°C
Catalog No. NC9990393 Supplier Enzo Life Sciences Supplier No. BMLSE1980025
Highly pure lysosomal protease. MW: ~27.5 kDa. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM sodium acetate, pH 5.0, containing 1mM EDTA. Source: Purified from human liver. UniProt: P07858. Long Term Storage: -80°C