Learn More
Trajan Scientific L-MARK Brand 25l, Syringe, GT, FN, 22s, PTFE, Pt. 3
Catalog No. 50635292
Encompass_Preferred
L-MARK Brand 25l, L-MARK Brand Syringe, FN, GT, 22s Gauge, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single, Syringe Adapter: PAL.SA025, Replacement Plunger: LMK.2636505 Made in Germany.
Catalog No. 50-635-292 Supplier Trajan Scientific Supplier No. LMK.2620517
L-MARK Brand 25l, L-MARK Brand Syringe, FN, GT, 22s Gauge, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single, Syringe Adapter: PAL.SA025, Replacement Plunger: LMK.2636505 Made in Germany.