Learn More
Ace Glass, Inc. 25 microliter gas-tight, fixed needle syringe
SDP
Catalog No. 50972628
Encompass
25 microliter gas-tight, fixed needle syringe. Syringe has a PTFE coated plunger with a precision PTFE tip for a leak-tight seal. Designed for highest performance in such applications as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions. Accuracy and reproducibility of plus or minus 1%.
Catalog No. 50-972-628 Supplier Ace Glass, Inc. Supplier No. 593103
25 microliter gas-tight, fixed needle syringe. Syringe has a PTFE coated plunger with a precision PTFE tip for a leak-tight seal. Designed for highest performance in such applications as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions. Accuracy and reproducibility of plus or minus 1%.