Learn More
Ace Glass, Inc. Congealing temperature 25x215mm test tube
SDP
Catalog No. NC9031347
Encompass
25x215mm congealing temperature test tube, part of the ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical.
Catalog No. NC9031347 Supplier Ace Glass, Inc. Supplier No. 654714
25x215mm congealing temperature test tube, part of the ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical.