Learn More
Bellco Glass, Inc. 25x25 Flask Tray, 4 1L pre-mounted clamps
SDP
Catalog No. 501946144
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 4 Dimensions: 25x25" Size: 1l
Catalog No. 50-194-6144 Supplier Bellco Glass, Inc. Supplier No. 774410004
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask positions: 4 Dimensions: 25x25" Size: 1l