Learn More
Bellco Glass, Inc. 25x25cm Flask Tray,250mL
SDP
Supplier:  Bellco Glass, Inc. 774425005
Encompass_Preferred
Pre-mounted stainless steel clamps for bellco Erlenmeyer flask. Flask tray fits on top of the standard tray platform. Includes clamps.. MADE IN THE USA Flask position: 5 Dimensions:25x25cm Size: 250ml
Catalog No. 50-194-6159