Learn More
MSC Value Collection Air Filter, MERV 8, 25x25x1 inch, 1 ea
Catalog No. 5000505226
Encompass
This pleated air filter is designed for use in heating and air conditioning units. It features a wireless pleated design with a MERV 8 rating, offering effective air filtration.
  • Wireless pleated design
  • MERV 8 filtration efficiency
  • Nominal size of 25 in x 25 in x 1 in
  • Synthetic media material
  • Sturdy moisture resistant beverage board frame
  • Airflow of 1300 CFM
  • Maximum velocity of 300 FPM
  • Complies with ASHRAE 52.2-2012 and UL 900 standards
  • Exclusive vForm self-supported pleats
Catalog No. 50-005-05226 Supplier MSC Supplier No. 03639549
This pleated air filter is designed for use in heating and air conditioning units. It features a wireless pleated design with a MERV 8 rating, offering effective air filtration.
  • Wireless pleated design
  • MERV 8 filtration efficiency
  • Nominal size of 25 in x 25 in x 1 in
  • Synthetic media material
  • Sturdy moisture resistant beverage board frame
  • Airflow of 1300 CFM
  • Maximum velocity of 300 FPM
  • Complies with ASHRAE 52.2-2012 and UL 900 standards
  • Exclusive vForm self-supported pleats