Learn More
Ace Glass, Inc. Adjusta-Chrom; column, 25mm ID x 300mm, complete with top& bottom adjusting plungers with PTFE seals and a 3 meter length of 2.0mm ID PTFE tubing
SDP
Supplier:  Ace Glass, Inc. 581515
Encompass_Preferred
25mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications. Supplied with top & bottom adjusting plungers with PTFE seals, 25-50mic glass disc, and a 3m length of 2mm I.D. PTFE tubing.
Catalog No. 50-972-144