Learn More
Ace Glass, Inc. Adjusta-Chrom; column, 25mm ID x 300mm, complete with top& bottom adjusting plungers with FETFE O-ring seals and a 3meter length of 2.0mm ID PTFE tubing
SDP
Catalog No. 50972145
Encompass_Preferred
25mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications. Supplied with top & bottom adjusting plungers with o-ring seals, 25-50mic glass disc, and a 3m length of 2mm I.D. PTFE tubing.
Catalog No. 50-972-145 Supplier Ace Glass, Inc. Supplier No. 581516
25mm I.D. x 300mm Adjusta-Chrom chromatographic column used for recycling, ascending or descending applications. Supplied with top & bottom adjusting plungers with o-ring seals, 25-50mic glass disc, and a 3m length of 2mm I.D. PTFE tubing.