Learn More
Glass Vials 8 Dram Clear 25x95 screw thread Vial w/22-400 PTFE Lined Cap. 30ml. Case of 72
Catalog No. NC0456635
Encompass_Preferred
8 Dram Clear 25x95 screw thread Vial w/22-400 PTFE Lined Cap. 30ml. Case of 72
Catalog No. NC0456635 Supplier Glass Vials Supplier No. V2595CTFE
8 Dram Clear 25x95 screw thread Vial w/22-400 PTFE Lined Cap. 30ml. Case of 72