Learn More
Medchemexpress LLC 26:0 Lyso PC-d4 | 1246303-15-0 | 95.6% | 639.92 | 1 MG
SDP
Catalog No. 5000362941
Encompass_Preferred
26:0 Lyso PC-d4 is deuterium labeled 26:0 Lyso PC. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-003-62941 Supplier Medchemexpress LLC Supplier No. HY146879S1MG
26:0 Lyso PC-d4 is deuterium labeled 26:0 Lyso PC. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only