Learn More
Ace Glass, Inc. 26.5 liter graduated cylindrical jar with side indents
SDP
Catalog No. 50978664
Encompass
26.5L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.
Catalog No. 50-978-664 Supplier Ace Glass, Inc. Supplier No. 623326
26.5L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.