Learn More
MSC North Chemical Resistant Gloves, Nitrile, Medium, Supported, Blue
Catalog No. 5000510040
Encompass
The product is a pair of chemical resistant gloves designed for various applications. These supported nitrile gloves are blue with a rough grip and feature a cotton jersey thermal lining, providing comfort and protection. They offer ANSI abrasion, cut, and puncture resistance, making them suitable for tasks requiring durability and chemical protection.
  • Lined and coated for added strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Made from nitrile material
  • Medium glove size
  • Rough grip surface
  • Cotton jersey thermal lining
  • Blue color
  • Knit cuff style
  • Insulated liner
  • Latex-free
  • Overall length of 26.00 in
  • Thickness range of less than 15 mil
Catalog No. 50-005-10040 Supplier MSC Supplier No. 56100175
The product is a pair of chemical resistant gloves designed for various applications. These supported nitrile gloves are blue with a rough grip and feature a cotton jersey thermal lining, providing comfort and protection. They offer ANSI abrasion, cut, and puncture resistance, making them suitable for tasks requiring durability and chemical protection.
  • Lined and coated for added strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Made from nitrile material
  • Medium glove size
  • Rough grip surface
  • Cotton jersey thermal lining
  • Blue color
  • Knit cuff style
  • Insulated liner
  • Latex-free
  • Overall length of 26.00 in
  • Thickness range of less than 15 mil