Learn More
Alkali Scientific 26' Cassette of 12mm Lab Tape, Clear with Black Print
SDP
Catalog No. 502389185
Encompass_Preferred
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This clear tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50˚C.
Catalog No. 50-238-9185 Supplier Alkali Scientific Supplier No. L901012CK
Proper identification of scientific samples is often a key objective of the research laboratory. The process of writing on sample tubes/containers can be messy and unreliable. It can also be difficult to find the time to access a computer and design a layout for expensive ink jet labels. This clear tape with black print is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50˚C.