Learn More
Abcam 2,6-Dichloroindophenol sodium salt, blue redo x indicator dye, 25G
Catalog No. 502704665
Encompass_Preferred
MW 290.08 g/mol. Blue redox indicator dye used for the determination of Vitamin C (ascorbic acid), which when Vitamin C is present reduces to a colorless hydroxy compound. May be used as an electron acceptor and to stain glutathione reductase electrophoresis gels. Also known as Tillman's reagent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4665 Supplier Abcam Supplier No. AB14638925G
MW 290.08 g/mol. Blue redox indicator dye used for the determination of Vitamin C (ascorbic acid), which when Vitamin C is present reduces to a colorless hydroxy compound. May be used as an electron acceptor and to stain glutathione reductase electrophoresis gels. Also known as Tillman's reagent.
The product is subject to the following: Abcam Restricted Use Statement