Learn More
Medchemexpress LLC HY-W010195 500mg Medchemexpress, 2,6-Dimethylquinoline CAS:877-43-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010195500MG
Encompass_Preferred
Medchemexpress, HY-W010195 500mg 2,6-Dimethylquinoline CAS:877-43-0 2,6-Dimethylquinoline, a nature constituent from the roots of Peucedantu praeruptorum, is a CYP1A2 inhibitor with an IC50 of 3.3 µM. 2,6-Dimethylquinoline also inhibits CYP2B6 activity with an IC50 of 480 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3285