Learn More
SPEX CERTIPREP LLC 3P 26 ELEMENT STD 100 UG/ML 100ML
Catalog No. NC0257183
Encompass_Preferred
(HPS by Zeptometrix) 26 element ICP quality control standard: Al, Sb, As, B, Ba, Be, Cd, Ca, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, K, Se, Si, Ag, Na, Tl, Titanium, V, Zn @ 100 ug/mL in 5% HNO3 + Tr HF
Catalog No. NC0257183 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCS26R100
(HPS by Zeptometrix) 26 element ICP quality control standard: Al, Sb, As, B, Ba, Be, Cd, Ca, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, K, Se, Si, Ag, Na, Tl, Titanium, V, Zn @ 100 ug/mL in 5% HNO3 + Tr HF